Gene Transcripts

Gene Symbol
MIMT1
Entrez Gene ID
100073347
Gene Name
MER1 repeat containing imprinted transcript 1 (non-protein coding)
Gene Aliases
LINC00067, MIM1, NCRNA00067
Chromosome Location
Chr. 19: 56840902 - 56848554 on Build GRCh38
UniGene ID
Hs.467337
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024059.1 2 984
GenBank mRNA EF110915.1 - 2 984

View Full Product Details