Gene Transcripts
- Gene Symbol
- TPI1P3
- Entrez Gene ID
- 728402
- Gene Name
- triosephosphate isomerase 1 pseudogene 3
- Gene Aliases
- Chromosome Location
- Chr. 6: 116038731 - 116039944 on Build GRCh38
- UniGene ID
- Hs.729953
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027338.1 |
|
1 |
1060 |
|
View Full Product Details