Gene Transcripts

Gene Symbol
SNHG5
Entrez Gene ID
387066
Gene Name
small nucleolar RNA host gene 5
Gene Aliases
C6orf160, LINC00044, NCRNA00044, U50HG, bA33E24.2
Chromosome Location
Chr. 6: 85677007 - 85678733 on Build GRCh38
UniGene ID
Hs.292457
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003038.2 4 203
GenBank mRNA BC009220.2 - 3 677
BC108291.1 - 3 695
CD384894.1 - 4 220
CD388674.1 - 4 220


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CD389879.1 - 4 226 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA CN368121.1 - 3 160 Chr. 6 - 85677470 - 85678185 [-]
GenBankmRNA CD173365.1 - 4 229 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA F28088.1 - 3 152 Chr. 6 - 85677829 - 85678177 [-]
GenBankmRNA F34856.1 - 200 Chr. - - [-]
GenBankmRNA DA482879.1 - 4 203 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA BF209379.1 - 4 205 Chr. 6 - 85677078 - 85678733 [-]
GenBankmRNA CD520230.1 - 4 206 Chr. 6 - 85677083 - 85678736 [-]
GenBankmRNA CD175099.1 - 4 225 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA BE715636.1 - 3 170 Chr. 6 - 85677108 - 85678173 [-]
GenBankmRNA BU532076.1 - 206 Chr. - - [-]
GenBankmRNA BE931917.1 - 3 344 Chr. 6 - 85677155 - 85678356 [-]
GenBankmRNA CD247680.1 - 4 223 Chr. 6 - 85677082 - 85678724 [-]
GenBankmRNA CD249906.1 - 4 209 Chr. 6 - 85677085 - 85678731 [-]
GenBankmRNA AW972834.1 - 297 Chr. - - [-]
GenBankmRNA BQ876776.1 - 183 Chr. - - [-]
GenBankmRNA CD389769.1 - 4 222 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD175213.1 - 4 219 Chr. 6 - 85677085 - 85678733 [-]
GenBankmRNA F30675.1 - 4 185 Chr. 6 - 85677439 - 85678715 [-]
GenBankmRNA F29466.1 - 3 171 Chr. 6 - 85677423 - 85678192 [-]
GenBankmRNA CD048591.1 - 225 Chr. - - [-]
GenBankmRNA BU536948.1 - 201 Chr. - - [-]
GenBankmRNA CN368113.1 - 3 182 Chr. 6 - 85677082 - 85678192 [-]
GenBankmRNA CD048947.1 - 226 Chr. - - [-]
GenBankmRNA BU500478.1 - 4 186 Chr. 6 - 85677082 - 85678717 [-]
GenBankmRNA BE392169.1 - 3 160 Chr. 6 - 85677079 - 85678185 [-]
GenBankmRNA CD388186.1 - 4 224 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA CD248605.1 - 4 221 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD177483.1 - 4 219 Chr. 6 - 85677085 - 85678733 [-]
GenBankmRNA CN368120.1 - 4 195 Chr. 6 - 85677082 - 85678717 [-]
GenBankmRNA CD175092.1 - 4 222 Chr. 6 - 85677082 - 85678724 [-]
GenBankmRNA BU588624.1 - 4 206 Chr. 6 - 85677082 - 85678736 [-]
GenBankmRNA CD249877.1 - 4 226 Chr. 6 - 85677079 - 85678725 [-]
GenBankmRNA CD385054.1 - 230 Chr. - - [-]
GenBankmRNA CD250044.1 - 4 231 Chr. 6 - 85677082 - 85678734 [-]
GenBankmRNA CD049899.1 - 221 Chr. - - [-]
GenBankmRNA CD248963.1 - 225 Chr. - - [-]
GenBankmRNA EL947347.1 - 4 191 Chr. 6 - 85677077 - 85678719 [-]
GenBankmRNA BM452606.1 - 3 162 Chr. 6 - 85677079 - 85678186 [-]
GenBankmRNA F24124.1 - 3 163 Chr. 6 - 85677795 - 85678188 [-]
GenBankmRNA F29578.1 - 3 160 Chr. 6 - 85677471 - 85678185 [-]
GenBankmRNA CD389793.1 - 224 Chr. - - [-]
GenBankmRNA BF966370.2 - 4 209 Chr. 6 - 85677082 - 85678737 [-]
GenBankmRNA BU526489.1 - 4 178 Chr. 6 - 85677082 - 85678709 [-]
GenBankmRNA N98867.1 - 3 199 Chr. 6 - 85677082 - 85678192 [-]
GenBankmRNA BG330935.1 - 4 194 Chr. 6 - 85677079 - 85678723 [-]
GenBankmRNA BM702833.1 - 3 581 Chr. 6 - 85677474 - 85678606 [-]
GenBankmRNA BF980513.1 - 180 Chr. - - [-]
GenBankmRNA W04663.1 - 186 Chr. - - [-]
GenBankmRNA BU588037.1 - 206 Chr. - - [-]
GenBankmRNA CB117414.1 - 4 182 Chr. 6 - 85677082 - 85678712 [-]
GenBankmRNA BG332327.1 - 187 Chr. - - [-]
GenBankmRNA CD672298.1 - 4 483 Chr. 6 - 85677470 - 85678722 [-]
GenBankmRNA BU191449.1 - 4 184 Chr. 6 - 85677082 - 85678715 [-]
GenBankmRNA BG705462.1 - 4 209 Chr. 6 - 85677082 - 85678737 [-]
GenBankmRNA AA308290.1 - 4 179 Chr. 6 - 85677083 - 85678709 [-]
GenBankmRNA AJ714332.1 - 4 196 Chr. 6 - 85677100 - 85678715 [-]
GenBankmRNA AW022412.1 - 3 222 Chr. 6 - 85677423 - 85678242 [-]
GenBankmRNA CD177664.1 - 4 223 Chr. 6 - 85677085 - 85678733 [-]
GenBankmRNA BU602968.1 - 207 Chr. - - [-]
GenBankmRNA CD172948.1 - 4 222 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA HY027691.1 - 4 202 Chr. 6 - 85677083 - 85678732 [-]
GenBankmRNA BU181781.1 - 180 Chr. - - [-]
GenBankmRNA CD174861.1 - 4 226 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA F21312.1 - 1 189 Chr. 1 - 206772417 - 206773743 [-]
GenBankmRNA BU941995.1 - 206 Chr. - - [-]
GenBankmRNA BG758157.1 - 4 188 Chr. 6 - 85677082 - 85678717 [-]
GenBankmRNA BQ896708.1 - 4 182 Chr. 6 - 85677082 - 85678713 [-]
GenBankmRNA CD174362.1 - 4 227 Chr. 6 - 85677082 - 85678734 [-]
GenBankmRNA BU536398.1 - 4 206 Chr. 6 - 85677082 - 85678736 [-]
GenBankmRNA CD172704.1 - 4 223 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA F25212.1 - 4 186 Chr. 6 - 85677105 - 85678716 [-]
GenBankmRNA BF663376.1 - 186 Chr. - - [-]
GenBankmRNA CD175429.1 - 4 221 Chr. 6 - 85677082 - 85678731 [-]
GenBankmRNA BU931690.1 - 3 146 Chr. 6 - 85677082 - 85678171 [-]
GenBankmRNA CD250358.1 - 1 229 Chr. 17 - 49339407 - 49339817 [-]
GenBankmRNA BG760195.1 - 4 187 Chr. 6 - 85677082 - 85678716 [-]
GenBankmRNA BE736061.1 - 3 168 Chr. 6 - 85677079 - 85678192 [-]
GenBankmRNA BF309660.1 - 3 175 Chr. 6 - 85677082 - 85678192 [-]
GenBankmRNA W58769.1 - 189 Chr. - - [-]
GenBankmRNA BU169141.1 - 4 185 Chr. 6 - 85677082 - 85678715 [-]
GenBankmRNA BM476100.1 - 192 Chr. - - [-]
GenBankmRNA HY104298.1 - 4 204 Chr. 6 - 85677083 - 85678732 [-]
GenBankmRNA BM469398.1 - 3 163 Chr. 6 - 85677079 - 85678188 [-]
GenBankmRNA BU600470.1 - 4 209 Chr. 6 - 85677077 - 85678736 [-]
GenBankmRNA F25999.1 - 4 187 Chr. 6 - 85677082 - 85678717 [-]
GenBankmRNA CD174641.1 - 4 225 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA CD246809.1 - 4 219 Chr. 6 - 85677085 - 85678731 [-]
GenBankmRNA CB996821.1 - 4 226 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA BG398388.1 - 181 Chr. - - [-]
GenBankmRNA BE257562.1 - 3 167 Chr. 6 - 85677081 - 85678192 [-]
GenBankmRNA CB110857.1 - 4 203 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD676465.1 - 3 246 Chr. 6 - 85677082 - 85678252 [-]
GenBankmRNA CD000147.1 - 207 Chr. - - [-]
GenBankmRNA BG754322.1 - 187 Chr. - - [-]
GenBankmRNA BU532819.1 - 4 206 Chr. 6 - 85677082 - 85678736 [-]
GenBankmRNA BG419815.1 - 4 196 Chr. 6 - 85677077 - 85678726 [-]
GenBankmRNA BG332320.1 - 4 187 Chr. 6 - 85677082 - 85678716 [-]
GenBankmRNA F20491.1 - 170 Chr. 17 - 9240179 - 9240205 [+]
GenBankmRNA HY067002.1 - 4 202 Chr. 6 - 85677082 - 85678732 [-]
GenBankmRNA CD174976.1 - 4 221 Chr. 6 - 85677085 - 85678733 [-]
GenBankmRNA F33656.1 - 157 Chr. - - [-]
GenBankmRNA CD387937.1 - 220 Chr. - - [-]
GenBankmRNA HX785786.1 - 4 555 Chr. 6 - 85677147 - 85678602 [-]
GenBankmRNA AV712595.2 - 4 203 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA BG759432.1 - 4 181 Chr. 6 - 85677082 - 85678710 [-]
GenBankmRNA CD250171.1 - 4 221 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA HY181073.1 - 4 203 Chr. 6 - 85677083 - 85678732 [-]
GenBankmRNA CD249518.1 - 4 229 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA HY031639.1 - 4 202 Chr. 6 - 85677084 - 85678732 [-]
GenBankmRNA F24362.1 - 3 159 Chr. 6 - 85677119 - 85678184 [-]
GenBankmRNA F19304.2 - 187 Chr. 11 - 8244443 - 8244457 [+]
GenBankmRNA CD177169.1 - 4 225 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA BM920267.1 - 187 Chr. - - [-]
GenBankmRNA CN368117.1 - 4 195 Chr. 6 - 85677007 - 85678717 [-]
GenBankmRNA EB388820.1 - 4 192 Chr. 6 - 85677082 - 85678722 [-]
GenBankmRNA BG333157.1 - 4 194 Chr. 6 - 85677084 - 85678723 [-]
GenBankmRNA CD385963.1 - 4 225 Chr. 6 - 85677082 - 85678731 [-]
GenBankmRNA BG107883.1 - 3 238 Chr. 6 - 85677082 - 85678262 [-]
GenBankmRNA F31772.1 - 3 160 Chr. 6 - 85677119 - 85678185 [-]
GenBankmRNA CD174499.1 - 4 225 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CF780557.1 - 4 186 Chr. 6 - 85677079 - 85678716 [-]
GenBankmRNA BU182289.1 - 4 189 Chr. 6 - 85677082 - 85678719 [-]
GenBankmRNA BQ230279.1 - 184 Chr. - - [-]
GenBankmRNA BM695213.1 - 4 185 Chr. 6 - 85677082 - 85678715 [-]
GenBankmRNA BU531826.1 - 4 206 Chr. 6 - 85677082 - 85678736 [-]
GenBankmRNA BG472221.1 - 697 Chr. - - [-]
GenBankmRNA DB449890.1 - 4 207 Chr. 6 - 85677082 - 85678736 [-]
GenBankmRNA BU954986.1 - 206 Chr. - - [-]
GenBankmRNA BF971639.1 - 187 Chr. - - [-]
GenBankmRNA BF678689.1 - 204 Chr. - - [-]
GenBankmRNA CD175168.1 - 4 225 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD639840.1 - 376 Chr. - - [-]
GenBankmRNA AA194358.1 - 3 176 Chr. 6 - 85677082 - 85678192 [-]
GenBankmRNA CD173866.1 - 4 222 Chr. 6 - 85677082 - 85678731 [-]
GenBankmRNA HX789156.1 - 4 555 Chr. 6 - 85677147 - 85678602 [-]
GenBankmRNA CD177114.1 - 4 225 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA BE715633.1 - 3 180 Chr. 6 - 85677108 - 85678192 [-]
GenBankmRNA BU532664.1 - 4 206 Chr. 6 - 85677085 - 85678731 [-]
GenBankmRNA CD247795.1 - 4 222 Chr. 6 - 85677085 - 85678733 [-]
GenBankmRNA BU845184.1 - 3 176 Chr. 6 - 85677082 - 85678192 [-]
GenBankmRNA F22587.1 - 164 Chr. - - [-]
GenBankmRNA CD175058.1 - 4 225 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA BU536013.1 - 4 206 Chr. 6 - 85677082 - 85678736 [-]
GenBankmRNA CD388557.1 - 4 222 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA CD175102.1 - 4 224 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA BF307294.1 - 181 Chr. - - [-]
GenBankmRNA CD390262.1 - 4 219 Chr. 6 - 85677085 - 85678731 [-]
GenBankmRNA CB962559.1 - 4 206 Chr. 6 - 85677079 - 85678736 [-]
GenBankmRNA F33998.1 - 182 Chr. - - [-]
GenBankmRNA BE902100.1 - 3 162 Chr. 6 - 85677085 - 85678187 [-]
GenBankmRNA BU196777.1 - 3 174 Chr. 6 - 85677082 - 85678183 [-]
GenBankmRNA BE901509.1 - 162 Chr. - - [-]
GenBankmRNA CB128964.1 - 4 203 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA BG759735.1 - 4 186 Chr. 6 - 85677082 - 85678716 [-]
GenBankmRNA HY168432.1 - 4 203 Chr. 6 - 85677083 - 85678732 [-]
GenBankmRNA BU855495.1 - 4 182 Chr. 6 - 85677082 - 85678716 [-]
GenBankmRNA BG697433.1 - 3 695 Chr. 6 - 85677426 - 85678720 [-]
GenBankmRNA BQ645788.1 - 4 184 Chr. 6 - 85677082 - 85678714 [-]
GenBankmRNA AA515848.1 - 4 193 Chr. 6 - 85677082 - 85678714 [-]
GenBankmRNA BM840935.1 - 4 225 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD248175.1 - 4 225 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA BQ897037.1 - 3 174 Chr. 6 - 85677082 - 85678188 [-]
GenBankmRNA CD174771.1 - 4 223 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD173592.1 - 4 228 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA AJ709757.1 - 4 191 Chr. 6 - 85677084 - 85678716 [-]
GenBankmRNA BU185686.1 - 4 184 Chr. 6 - 85677082 - 85678715 [-]
GenBankmRNA BF218089.1 - 4 207 Chr. 6 - 85677079 - 85678736 [-]
GenBankmRNA CD173436.1 - 4 225 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD175427.1 - 4 225 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA AA385186.1 - 4 181 Chr. 6 - 85677169 - 85678711 [-]
GenBankmRNA CD177627.1 - 4 230 Chr. 6 - 85677082 - 85678734 [-]
GenBankmRNA F27384.1 - 3 160 Chr. 6 - 85677437 - 85678185 [-]
GenBankmRNA BE513854.1 - 3 163 Chr. 6 - 85677081 - 85678188 [-]
GenBankmRNA F19218.2 - 170 Chr. 1 - 203128824 - 203128850 [-]
GenBankmRNA BE256443.1 - 169 Chr. - - [-]
GenBankmRNA BI090148.1 - 187 Chr. - - [-]
GenBankmRNA CD175562.1 - 4 225 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD176074.1 - 4 229 Chr. 6 - 85677085 - 85678734 [-]
GenBankmRNA AJ574636.1 - 4 194 Chr. 6 - 85677087 - 85678718 [-]
GenBankmRNA CD175083.1 - 4 221 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA CD048516.1 - 260 Chr. - - [-]
GenBankmRNA CD640064.1 - 183 Chr. - - [-]
GenBankmRNA BE900923.1 - 3 167 Chr. 6 - 85677084 - 85678192 [-]
GenBankmRNA HY101068.1 - 4 202 Chr. 6 - 85677079 - 85678732 [-]
GenBankmRNA CD175206.1 - 4 232 Chr. 6 - 85677077 - 85678733 [-]
GenBankmRNA BG754036.1 - 4 186 Chr. 6 - 85677082 - 85678716 [-]
GenBankmRNA AA314904.1 - 3 164 Chr. 6 - 85677123 - 85678189 [-]
GenBankmRNA CD172637.1 - 4 221 Chr. 6 - 85677007 - 85678733 [-]
GenBankmRNA CD177343.1 - 4 226 Chr. 6 - 85677085 - 85678732 [-]
GenBankmRNA BM763888.1 - 4 203 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA BU944864.1 - 3 559 Chr. 6 - 85677082 - 85678584 [-]
GenBankmRNA CD675327.1 - 4 199 Chr. 6 - 85677082 - 85678720 [-]
GenBankmRNA AA313726.1 - 3 164 Chr. 6 - 85677077 - 85678189 [-]
GenBankmRNA CD173371.1 - 4 229 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD248801.1 - 4 221 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD248278.1 - 4 228 Chr. 6 - 85677077 - 85678733 [-]
GenBankmRNA BE562519.1 - 168 Chr. - - [-]
GenBankmRNA CD388259.1 - 4 226 Chr. 6 - 85677075 - 85678732 [-]
GenBankmRNA BG339050.1 - 184 Chr. - - [-]
GenBankmRNA CD390099.1 - 4 218 Chr. 6 - 85677083 - 85678721 [-]
GenBankmRNA N55638.1 - 3 199 Chr. 6 - 85677127 - 85678192 [-]
GenBankmRNA BM772443.1 - 4 203 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA BE812115.1 - 3 185 Chr. 6 - 85677119 - 85678192 [-]
GenBankmRNA F24856.1 - 4 182 Chr. 6 - 85677133 - 85678712 [-]
GenBankmRNA BU536630.1 - 4 206 Chr. 6 - 85677079 - 85678736 [-]
GenBankmRNA CD176142.1 - 4 193 Chr. 6 - 85677082 - 85678723 [-]
GenBankmRNA CN368115.1 - 4 200 Chr. 6 - 85677087 - 85678724 [-]
GenBankmRNA BG424974.1 - 4 196 Chr. 6 - 85677084 - 85678726 [-]
GenBankmRNA CB956756.1 - 4 203 Chr. 6 - 85677079 - 85678736 [-]
GenBankmRNA BG472840.1 - 4 187 Chr. 6 - 85677114 - 85678716 [-]
GenBankmRNA F36060.1 - 140 Chr. - - [-]
GenBankmRNA CD172639.1 - 4 224 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD389175.1 - 4 223 Chr. 6 - 85677082 - 85678732 [-]
GenBankmRNA AJ707924.1 - 4 221 Chr. 6 - 85677084 - 85678716 [-]
GenBankmRNA CD248034.1 - 5 228 Chr. 6 - 85677079 - 86545552 [-]
GenBankmRNA AB017710.3 - 1 1515 Chr. 6 - 85677007 - 85679361 [-]
GenBankmRNA BM006651.1 - 4 187 Chr. 6 - 85677082 - 85678716 [-]
GenBankmRNA CD248379.1 - 4 228 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD638710.1 - 4 206 Chr. 6 - 85677079 - 85678736 [-]
GenBankmRNA CD390234.1 - 4 228 Chr. 6 - 85677079 - 85678736 [-]
GenBankmRNA BG685417.1 - 4 181 Chr. 6 - 85677082 - 85678710 [-]
GenBankmRNA CD247900.1 - 4 223 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD173118.1 - 4 220 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA HY103231.1 - 4 201 Chr. 6 - 85677082 - 85678731 [-]
GenBankmRNA F37437.1 - 186 Chr. - - [-]
GenBankmRNA AL525144.3 - 3 699 Chr. 6 - 85677103 - 85678710 [-]
GenBankmRNA T57445.1 - 175 Chr. - - [-]
GenBankmRNA F31547.1 - 3 170 Chr. 6 - 85677831 - 85678192 [-]
GenBankmRNA CB117807.1 - 4 202 Chr. 6 - 85677472 - 85678731 [-]
GenBankmRNA F27612.1 - 3 164 Chr. 6 - 85677443 - 85678189 [-]
GenBankmRNA CD175900.1 - 4 223 Chr. 6 - 85677079 - 85678731 [-]
GenBankmRNA CN368114.1 - 4 202 Chr. 6 - 85677087 - 85678720 [-]
GenBankmRNA BF974828.1 - 4 181 Chr. 6 - 85677082 - 85678710 [-]
GenBankmRNA CD174275.1 - 4 225 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA BM841447.1 - 4 224 Chr. 6 - 85677134 - 85678733 [-]
GenBankmRNA BQ935537.1 - 3 206 Chr. 6 - 85677075 - 85678191 [-]
GenBankmRNA AA401334.1 - 3 169 Chr. 6 - 85677047 - 85678192 [-]
GenBankmRNA CD173175.1 - 4 220 Chr. 6 - 85677085 - 85678733 [-]
GenBankmRNA AA315506.1 - 4 181 Chr. 6 - 85677085 - 85678711 [-]
GenBankmRNA DW432544.1 - 4 220 Chr. 6 - 85677800 - 85678736 [-]
GenBankmRNA CD174931.1 - 4 220 Chr. 6 - 85677082 - 85678733 [-]

View Full Product Details