Gene Transcripts

Gene Symbol
HULC
Entrez Gene ID
728655
Gene Name
hepatocellular carcinoma up-regulated long non-coding RNA
Gene Aliases
HCCAT1, LINC00078, NCRNA00078
Chromosome Location
Chr. 6: 8652209 - 8653846 on Build GRCh38
UniGene ID
Hs.214343
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_004855.2 1 116
GenBank mRNA AY914050.1 - 1 116


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BX116704.1 - 1 78 Chr. 6 - 8652247 - 8653846 [+]

View Full Product Details