Gene Transcripts
- Gene Symbol
- HULC
- Entrez Gene ID
- 728655
- Gene Name
- hepatocellular carcinoma up-regulated long non-coding RNA
- Gene Aliases
- HCCAT1, LINC00078, NCRNA00078
- Chromosome Location
- Chr. 6: 8652209 - 8653846 on Build GRCh38
- UniGene ID
- Hs.214343
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_004855.2 |
|
1 |
67 |
|
GenBank mRNA |
AY914050.1 |
- |
1 |
67 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX116704.1 |
- |
1 |
29 |
|
Chr. 6 - 8652247 - 8653846 [+] |
View Full Product Details