Gene Transcripts

Gene Symbol
SLC4A3
Entrez Gene ID
6508
Gene Name
solute carrier family 4 member 3
Gene Aliases
AE3, CAE3/BAE3, SLC2C
Chromosome Location
Chr. 2: 219627570 - 219641980 on Build GRCh38
UniGene ID
Hs.1176
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005246790.4 XP_005246847.1 1 95
GenBank mRNA U05597.1 - 1 49

View Full Product Details