Gene Transcripts
- Gene Symbol
- KRTAP4-6
- Entrez Gene ID
- 81871
- Gene Name
- keratin associated protein 4-6
- Gene Aliases
- KAP4.15, KAP4.6, KRTAP4-15, KRTAP4.15
- Chromosome Location
- Chr. 17: 41139433 - 41140487 on Build GRCh38
- UniGene ID
- Hs.307014
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AJ406938.1 |
- |
1 |
640 |
|
AJ406945.1 |
- |
1 |
905 |
|
View Full Product Details