Gene Transcripts

Gene Symbol
FUBP1
Entrez Gene ID
8880
Gene Name
far upstream element binding protein 1
Gene Aliases
FBP, FUBP, hDH V
Chromosome Location
Chr. 1: 77944055 - 77979494 on Build GRCh38
UniGene ID
Hs.567380
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011542391.2 XP_011540693.1 22 2294
XM_011542392.2 XP_011540694.1 22 2325
XM_011542393.2 XP_011540695.1 21 2265
XM_017002739.1 XP_016858228.1 23 2316
XM_017002740.1 XP_016858229.1 21 4157
XM_017002741.1 XP_016858230.1 22 2309
XM_017002742.1 XP_016858231.1 21 5410
XM_017002743.1 XP_016858232.1 20 4129
XM_017002744.1 XP_016858233.1 20 5351
GenBank mRNA AL049951.1 - 1 306


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BX111567.1 - 23 Chr. - - [-]

View Full Product Details