Gene Transcripts

Gene Symbol
SLC25A28
Entrez Gene ID
81894
Gene Name
solute carrier family 25 member 28
Gene Aliases
MFRN2, MRS3/4, MRS4L, NPD016
Chromosome Location
Chr. 10: 99610518 - 99659329 on Build GRCh38
UniGene ID
Hs.403790
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011540240.1 XP_011538542.1 1 897
XM_017016741.1 XP_016872230.1 1 897
GenBank mRNA AK094405.1 - 1 898

View Full Product Details