Gene Transcripts
- Gene Symbol
- LCLAT1
- Entrez Gene ID
- 253558
- Gene Name
- lysocardiolipin acyltransferase 1
- Gene Aliases
- 1AGPAT8, AGPAT8, ALCAT1, HSRG1849, LYCAT, UNQ1849
- Chromosome Location
- Chr. 2: 30447226 - 30644225 on Build GRCh38
- UniGene ID
- Hs.468048
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK095284.1 |
- |
7 |
1677 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX647670.1 |
- |
1 |
4056 |
|
Chr. 6 - 73309524 - 73314589 [+] |
View Full Product Details