Gene Transcripts
- Gene Symbol
- LOC105373495
- Entrez Gene ID
- 105373495
- Gene Name
- uncharacterized LOC105373495
- Gene Aliases
- Chromosome Location
- Chr. 2: 96307263 - 96321893 on Build GRCh38
- UniGene ID
- Hs.379222
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC015425.1 |
- |
1 |
265 |
|
BC053735.1 |
- |
1 |
290 |
|
- Gene Symbol
- SNRNP200
- Entrez Gene ID
- 23020
- Gene Name
- small nuclear ribonucleoprotein U5 subunit 200
- Gene Aliases
- ASCC3L1, BRR2, HELIC2, RP33, U5-200KD
- Chromosome Location
- Chr. 2: 96274336 - 96321893 on Build GRCh38
- UniGene ID
- Hs.246112
- Species
- Homo sapiens
View Full Product Details