Gene Transcripts

Gene Symbol
ZMYND8
Entrez Gene ID
23613
Gene Name
zinc finger MYND-type containing 8
Gene Aliases
PRKCBP1, PRO2893, RACK7
Chromosome Location
Chr. 20: 47209214 - 47357780 on Build GRCh38
UniGene ID
Hs.446240
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006723760.3 XP_006723823.1 1 36
GenBank mRNA AF144233.1 - 1 756

View Full Product Details