Gene Transcripts
- Gene Symbol
- FAM63B
- Entrez Gene ID
- 54629
- Gene Name
- family with sequence similarity 63 member B
- Gene Aliases
- Chromosome Location
- Chr. 15: 58771192 - 58861900 on Build GRCh38
- UniGene ID
- Hs.591122
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA365499.1 |
- |
1 |
23 |
|
Chr. 15 - 58860052 - 58860627 [+] |
GenBankmRNA |
DA264557.1 |
- |
1 |
24 |
|
Chr. 15 - 58860051 - 58860627 [+] |
GenBankmRNA |
CR739799.1 |
- |
1 |
228 |
|
Chr. 15 - 58859852 - 58860472 [+] |
GenBankmRNA |
DA071804.1 |
- |
1 |
24 |
|
Chr. 15 - 58860051 - 58860619 [+] |
GenBankmRNA |
CN360071.1 |
- |
|
143 |
|
Chr. - - [-] |
View Full Product Details