Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033104.3 | NP_149095.2 | 6 | 6964 | |
XM_006720279.1 | XP_006720342.1 | 6 | 4681 | ||
XM_011537242.1 | XP_011535544.1 | 7 | 4852 | ||
GenBank mRNA | BX648249.1 | - | 1 | 638 |