Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_152573.3 | NP_689786.2 | 17 | 4965 | |
XM_005251730.3 | XP_005251787.1 | 16 | 4945 | ||
XM_005251731.3 | XP_005251788.1 | 17 | 5456 | ||
GenBank mRNA | AK056882.1 | - | 1 | 2241 |