Gene Transcripts
- Gene Symbol
- SCARB2
- Entrez Gene ID
- 950
- Gene Name
- scavenger receptor class B member 2
- Gene Aliases
- AMRF, CD36L2, EPM4, HLGP85, LGP85, LIMP-2, LIMPII, SR-BII
- Chromosome Location
- Chr. 4: 76158737 - 76213899 on Build GRCh38
- UniGene ID
- Hs.349656
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK022432.1 |
- |
1 |
1056 |
|
View Full Product Details