Gene Transcripts

Gene Symbol
Senp1
Entrez Gene ID
223870
Gene Name
SUMO1/sentrin specific peptidase 1
Gene Aliases
2310046A20Rik, D15Ertd528e, E330036L07Rik, suPr-2
Chromosome Location
Chr. 15: 98038744 - 98093569 on Build GRCm38
UniGene ID
Mm.384023
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK009838.1 - 1 2256


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CO809827.1 - 1 276 Chr. 15 - 98038758 - 98039198 [-]
GenBankmRNA BI083191.1 - 1 228 Chr. 15 - 98038765 - 98039162 [-]

View Full Product Details