Gene Transcripts
- Gene Symbol
- Rapgef6
- Entrez Gene ID
- 192786
- Gene Name
- Rap guanine nucleotide exchange factor (GEF) 6
- Gene Aliases
- A530068K01, AI844632, BB085664, C030018K18Rik, PDZ-GEF2, Pdzgef2, RA-GEF-2, Ragef2, mKIAA4052
- Chromosome Location
- Chr. 11: 54521754 - 54699286 on Build GRCm38
- UniGene ID
- Mm.254404
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK021094.1 |
- |
1 |
344 |
|
AK038235.1 |
- |
1 |
953 |
|
View Full Product Details