Gene Transcripts
- Gene Symbol
- Med14
- Entrez Gene ID
- 26896
- Gene Name
- mediator complex subunit 14
- Gene Aliases
- 9930001L01Rik, AU041628, Crsp2, ENSMUSG00000073278, Gm641, ORF1, Trap170
- Chromosome Location
- Chr. X: 12675368 - 12762042 on Build GRCm38
- UniGene ID
- Mm.17616
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK036758.1 |
- |
1 |
1042 |
|
View Full Product Details