Gene Transcripts
- Gene Symbol
- Pdzd8
- Entrez Gene ID
- 107368
- Gene Name
- PDZ domain containing 8
- Gene Aliases
- A630041P07Rik, AV329468, AW228944, Pdzk8
- Chromosome Location
- Chr. 19: 59296084 - 59345780 on Build GRCm38
- UniGene ID
- Mm.268797
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK088487.1 |
- |
|
3626 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY762603.1 |
- |
1 |
611 |
|
Chr. 19 - 59293231 - 59293890 [-] |
View Full Product Details