Gene Transcripts
- Gene Symbol
- Numb
- Entrez Gene ID
- 18222
- Gene Name
- numb gene homolog (Drosophila)
- Gene Aliases
- Nb
- Chromosome Location
- Chr. 12: 83794034 - 83921934 on Build GRCm38
- UniGene ID
- Mm.4390
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK169548.1 |
- |
1 |
1851 |
|
View Full Product Details