Gene Transcripts
- Gene Symbol
- DLX6-AS1
- Entrez Gene ID
- 285987
- Gene Name
- DLX6 antisense RNA 1
- Gene Aliases
- DLX6-AS, DLX6AS, Evf-2, NCRNA00212
- Chromosome Location
- Chr. 7: 96968515 - 97014065 on Build GRCh38
- UniGene ID
- Hs.34969
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK055423.1 |
- |
1 |
2013 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000452769.3 |
- |
2 |
3623 |
|
Chr. 7 - 96965528 - 97014064 [-] |
GENCODE |
ENST00000431497.6 |
- |
2 |
4273 |
|
Chr. 7 - 96965528 - 97004298 [-] |
GENCODE |
ENST00000430027.3 |
- |
3 |
4869 |
|
Chr. 7 - 96955141 - 97014065 [-] |
View Full Product Details