Gene Transcripts
- Gene Symbol
- USP2-AS1
- Entrez Gene ID
- 100499227
- Gene Name
- USP2 antisense RNA 1 (head to head)
- Gene Aliases
- THY1-AS1
- Chromosome Location
- Chr. 11: 119381778 - 119499233 on Build GRCh38
- UniGene ID
- Hs.662365
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK097820.1 |
- |
4 |
490 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000500970.5 |
- |
4 |
490 |
|
Chr. 11 - 119381810 - 119526664 [+] |
View Full Product Details