Gene Transcripts

Gene Symbol
UBR5
Entrez Gene ID
51366
Gene Name
ubiquitin protein ligase E3 component n-recognin 5
Gene Aliases
DD5, EDD, EDD1, HYD
Chromosome Location
Chr. 8: 102252273 - 102412719 on Build GRCh38
UniGene ID
Hs.492445
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BX489192.1 - 1 277

Gene Symbol
UBR5-AS1
Entrez Gene ID
101927221
Gene Name
UBR5 antisense RNA 1
Gene Aliases
Chromosome Location
Chr. 8: 102239451 - 102252174 on Build GRCh38
UniGene ID
Hs.492445
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK095151.1 - 2 1248


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000499653.1 - 2 1248 Chr. 8 - 102243609 - 102253333 [+]

View Full Product Details