Gene Transcripts

Gene Symbol
TRIM55
Entrez Gene ID
84675
Gene Name
tripartite motif containing 55
Gene Aliases
MURF-2, RNF29, muRF2
Chromosome Location
Chr. 8: 66112667 - 66175485 on Build GRCh38
UniGene ID
Hs.85524
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005251316.4 XP_005251373.1 1 263
XM_011517613.2 XP_011515915.1 1 263
XM_011517615.2 XP_011515917.1 1 263
XM_011517616.2 XP_011515918.1 1 263
XM_011517617.2 XP_011515919.1 1 384


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000517961.3 - 1 263 Chr. 8 - 66113287 - 66115205 [+]

View Full Product Details