Gene Transcripts

Gene Symbol
PCAT1
Entrez Gene ID
100750225
Gene Name
prostate cancer associated transcript 1 (non-protein coding)
Gene Aliases
PCA1, PCAT-1
Chromosome Location
Chr. 8: 127013154 - 127021014 on Build GRCh38
UniGene ID
Hs.571530
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_045262.1 1 235
GenBank mRNA HQ605084.1 - 1 235


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000519319.1 - 5 409 Chr. 8 - 126847055 - 127013713 [+]
GENCODE ENST00000561978.1 - 1 235 Chr. 8 - 127013154 - 127021014 [+]
GenBankmRNA CR741388.1 - 5 409 Chr. 8 - 126847055 - 127013705 [+]

View Full Product Details