Gene Transcripts
- Gene Symbol
- Snhg5
- Entrez Gene ID
- 72655
- Gene Name
- small nucleolar RNA host gene 5
- Gene Aliases
- 2810026P18Rik, 4933428I09Rik, mU50HG-b
- Chromosome Location
- Chr. 9: 88521052 - 88522897 on Build GRCm38
- UniGene ID
- Mm.21798
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY711938.1 |
- |
1 |
17 |
|
Chr. 9 - 88521073 - 88522890 [-] |
GenBankmRNA |
BU937714.1 |
- |
1 |
59 |
|
Chr. 9 - 88521050 - 88522929 [-] |
GenBankmRNA |
CA467755.1 |
- |
1 |
52 |
|
Chr. 9 - 88521503 - 88522888 [-] |
View Full Product Details