Gene Transcripts

Gene Symbol
Mtmr11
Entrez Gene ID
194126
Gene Name
myotubularin related protein 11
Gene Aliases
AI035306, BC051083, Cra
Chromosome Location
Chr. 3: 96161946 - 96171718 on Build GRCm38
UniGene ID
Mm.268536
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181409.3 NP_852074.2 8 937
GenBank mRNA AK132375.1 - 8 904
BC051083.1 - 8 870 6486043

View Full Product Details