Gene Transcripts

Gene Symbol
Nhs
Entrez Gene ID
195727
Gene Name
Nance-Horan syndrome (human)
Gene Aliases
Gm389, Gm48, Nhs1, Xcat
Chromosome Location
Chr. X: 161833290 - 162159441 on Build GRCm38
UniGene ID
Mm.336313
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081052.2 NP_001074521.1 8 4582
NM_001290526.1 NP_001277455.1 9 4645
GenBank mRNA BC147632.1 - 8 4582
BC158016.1 - 9 4645 9007322
BC172134.1 - 9 4645 9054804

View Full Product Details