Gene Transcripts
- Gene Symbol
- Trav3-4
- Entrez Gene ID
- 211027
- Gene Name
- T cell receptor alpha variable 3-4
- Gene Aliases
- A730076H11Rik, Gm28, Gm79
- Chromosome Location
- Chr. 14: 53777071 - 53777576 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK043253.1 |
- |
|
616 |
|
View Full Product Details