Gene Transcripts

Gene Symbol
Tapbpl
Entrez Gene ID
213233
Gene Name
TAP binding protein-like
Gene Aliases
BC017613, TAPBPL-R, Tapbplr
Chromosome Location
Chr. 6: 125223922 - 125231860 on Build GRCm38
UniGene ID
Mm.296902
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145391.2 NP_663366.2 4 1099
GenBank mRNA BC017613.1 - 4 1099
BC094656.1 - 4 1069

View Full Product Details