Gene Transcripts
- Gene Symbol
- Rfesd
- Entrez Gene ID
- 218341
- Gene Name
- Rieske (Fe-S) domain containing
- Gene Aliases
- AI256775, D030068K09
- Chromosome Location
- Chr. 13: 76001535 - 76018712 on Build GRCm38
- UniGene ID
- Mm.266515
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001131068.1 |
NP_001124540.1
|
2, 3 |
761 |
|
NM_001131069.1 |
NP_001124541.1
|
2, 3 |
224 |
|
NM_178916.5 |
NP_849247.3
|
2, 3 |
1211 |
|
GenBank mRNA |
AK029602.1 |
- |
2, 3 |
1036 |
|
AK040255.1 |
- |
2, 3 |
716 |
|
AK083705.1 |
- |
2, 3 |
1212 |
|
AK163574.1 |
- |
2, 3 |
624 |
|
BC030381.1 |
- |
2, 3 |
836 |
5375753 |
CD769672.1 |
- |
2, 3 |
228 |
|
CN839985.1 |
- |
2, 3 |
234 |
|
View Full Product Details