Gene Transcripts
- Gene Symbol
- Arl15
- Entrez Gene ID
- 218639
- Gene Name
- ADP-ribosylation factor-like 15
- Gene Aliases
- A430036I03, Arfrp2, C230032K13Rik
- Chromosome Location
- Chr. 13: 113794506 - 114157462 on Build GRCm38
- UniGene ID
- Mm.79837
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172595.4 |
NP_766183.1
|
3, 4 |
403 |
|
GenBank mRNA |
AK039965.1 |
- |
3, 4 |
367 |
|
AK048736.1 |
- |
3, 4 |
286 |
|
AK050979.1 |
- |
3, 4 |
318 |
|
AK052164.1 |
- |
3, 4 |
352 |
|
AK053953.1 |
- |
4, 5 |
470 |
|
BC120664.1 |
- |
3, 4 |
289 |
40129587 |
BC120666.1 |
- |
3, 4 |
289 |
40129589 |
BC144993.1 |
- |
2, 3 |
229 |
9053541 |
CJ063085.1 |
- |
3, 4 |
403 |
|
CJ066332.1 |
- |
3, 4 |
387 |
|
View Full Product Details