Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177630.3 | NP_808298.2 | 1 | 1192 | |
GenBank mRNA | AK083881.1 | - | 1 | 1154 | |
BC035491.1 | - | 1 | 248 | ||
BC058638.1 | - | 1 | 861 | 6418256 | |
BC094559.1 | - | 1 | 1192 | 6826763 |