Gene Transcripts

Gene Symbol
Klhl14
Entrez Gene ID
225266
Gene Name
kelch-like 14
Gene Aliases
6330403N15Rik, AI785250, printor
Chromosome Location
Chr. 18: 21550377 - 21672932 on Build GRCm38
UniGene ID
Mm.436836
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081403.1 NP_001074872.1 5 1306
GenBank mRNA AK173163.1 - 5 1020

View Full Product Details