Gene Transcripts
- Gene Symbol
- Ndufs8
- Entrez Gene ID
- 225887
- Gene Name
- NADH dehydrogenase (ubiquinone) Fe-S protein 8
- Gene Aliases
- BC021616, CI-23kD, TYKY
- Chromosome Location
- Chr. 19: 3908863 - 3912774 on Build GRCm38
- UniGene ID
- Mm.44227
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271443.1 |
NP_001258372.1
|
3, 4 |
173 |
|
NM_001271444.1 |
NP_001258373.1
|
3, 4 |
413 |
|
NM_144870.5 |
NP_659119.2
|
3, 4 |
416 |
|
GenBank mRNA |
AK135069.1 |
- |
3, 4 |
174 |
|
AK149674.1 |
- |
3, 4 |
374 |
|
AK153182.1 |
- |
3, 4 |
360 |
|
AK153900.1 |
- |
3, 4 |
150 |
|
AK164901.1 |
- |
3, 4 |
150 |
|
AK167907.1 |
- |
3, 4 |
151 |
|
BC021616.1 |
- |
3, 4 |
134 |
5132866 |
BC086766.1 |
- |
3, 4 |
127 |
6820786 |
View Full Product Details