Gene Transcripts

Gene Symbol
Cyp2a22
Entrez Gene ID
233005
Gene Name
cytochrome P450, family 2, subfamily a, polypeptide 22
Gene Aliases
EG233005
Chromosome Location
Chr. 7: 26931631 - 26939412 on Build GRCm38
UniGene ID
Mm.291574
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101467.1 NP_001094937.1 8 1219
GenBank mRNA AK149509.1 - 7 2439
BC151116.1 - 8 1213 9088029
BC158034.1 - 8 1213 9007337

View Full Product Details