Gene Transcripts

Gene Symbol
Tmem45b
Entrez Gene ID
235135
Gene Name
transmembrane protein 45b
Gene Aliases
BC018222
Chromosome Location
Chr. 9: 31426196 - 31464238 on Build GRCm38
UniGene ID
Mm.390134
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144936.1 NP_659185.1 2 229
GenBank mRNA AK079262.1 - 2 215
AK144325.1 - 2 454
BC018222.1 - 2 185 4221959
BC026654.1 - 2 229 4988382

View Full Product Details