Gene Transcripts

Gene Symbol
POLR2I
Entrez Gene ID
5438
Gene Name
RNA polymerase II subunit I
Gene Aliases
RPB9, hRPB14.5
Chromosome Location
Chr. 19: 36113709 - 36115304 on Build GRCh38
UniGene ID
Hs.47062
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006233.4 NP_006224.1 3 611
GenBank mRNA AK312206.1 - 3 339
BC017112.1 - 3 473 3608402
BC067794.1 - 3 653 5285070

View Full Product Details