Gene Transcripts

Gene Symbol
PON1
Entrez Gene ID
5444
Gene Name
paraoxonase 1
Gene Aliases
ESA, MVCD5, PON
Chromosome Location
Chr. 7: 95298357 - 95324572 on Build GRCh38
UniGene ID
Hs.370995
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000446.5 NP_000437.3 7 855
GenBank mRNA AK314027.1 - 7 781
BC074719.2 - 7 787 30915169
D84371.1 - 7 812
M63012.1 - 7 767
M63013.1 - 7 706
M63014.1 - 6 672
S64615.1 - 7 767
U53784.1 - 7 812
Z70723.1 - 7 812

View Full Product Details