Gene Transcripts

Gene Symbol
Tspyl3
Entrez Gene ID
241732
Gene Name
TSPY-like 3
Gene Aliases
AW212607, Gm36
Chromosome Location
Chr. 2: 153222370 - 153225441 on Build GRCm38
UniGene ID
Mm.277243
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198617.2 NP_941019.2 1 677
GenBank mRNA AK134522.1 - 1 674
AK148882.1 - 1 622
AK167722.1 - 1 658
BC058340.1 - 1 619 6590443

View Full Product Details