Gene Transcripts

Gene Symbol
Sdr16c5
Entrez Gene ID
242285
Gene Name
short chain dehydrogenase/reductase family 16C, member 5
Gene Aliases
Rdhe2, Scdr9
Chromosome Location
Chr. 4: 3995942 - 4019663 on Build GRCm38
UniGene ID
Mm.98096
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181989.1 NP_871789.1 7 1091
GenBank mRNA AK136508.1 - 6 1062
AY277588.1 - 7 1091
BC104134.1 - 7 1010 40045268
BC104135.1 - 7 1010 40045269

View Full Product Details