Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290395.1 | NP_001277324.1 | 3 | 519 | |
NM_177746.4 | NP_808414.2 | 2 | 149 | ||
GenBank mRNA | AK034920.1 | - | 3 | 520 | |
AY611031.1 | - | 2 | 124 | ||
AY611032.1 | - | 2 | 149 | ||
BC138085.1 | - | 2 | 117 | 8861104 | |
BC138086.1 | - | 2 | 117 | 8861105 |