Gene Transcripts

Gene Symbol
Nlrc3
Entrez Gene ID
268857
Gene Name
NLR family, CARD domain containing 3
Gene Aliases
CLR16.2, D230007K08Rik, mFLJ00348
Chromosome Location
Chr. 16: 3943807 - 3979032 on Build GRCm38
UniGene ID
Mm.215994
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081280.1 NP_001074749.1 13, 14 3131
NM_175547.3 NP_780756.1 10, 11 1054
GenBank mRNA AK079766.1 - 10, 11 978
AK084199.1 - 10, 11 968
AK171971.1 - 12, 13 4144
BC130223.1 - 10, 11 2520
BC152543.1 - 10, 11 2529

View Full Product Details