Gene Transcripts

Gene Symbol
Spef2
Entrez Gene ID
320277
Gene Name
sperm flagellar 2
Gene Aliases
C230086A09Rik
Chromosome Location
Chr. 15: 9578193 - 9749044 on Build GRCm38
UniGene ID
Mm.127822
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK048961.1 - 14, 15 2049
BC132098.1 - 14, 15 1986 40130375
BC132288.1 - 14, 15 1986 40130565
HQ856050.1 - 15, 16 2120

View Full Product Details