Gene Transcripts

Gene Symbol
Mcc
Entrez Gene ID
328949
Gene Name
mutated in colorectal cancers
Gene Aliases
D18Ertd451e, E330037C19
Chromosome Location
Chr. 18: 44425060 - 44812182 on Build GRCm38
UniGene ID
Mm.312511
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085373.1 NP_001078842.1 15 2255
NM_001085374.1 NP_001078843.1 13 2223
GenBank mRNA AK054528.1 - 13 2226
AK158266.1 - 15 2256

View Full Product Details