Gene Transcripts

Gene Symbol
Lemd3
Entrez Gene ID
380664
Gene Name
LEM domain containing 3
Gene Aliases
AI316861, Man1
Chromosome Location
Chr. 10: 120923411 - 120979949 on Build GRCm38
UniGene ID
Mm.339371
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081193.2 NP_001074662.2 1 1500
GenBank mRNA BC082610.1 - 1 198

View Full Product Details