Gene Transcripts

Gene Symbol
Poc1b
Entrez Gene ID
382406
Gene Name
POC1 centriolar protein homolog B (Chlamydomonas)
Gene Aliases
4933430F16Rik, Wdr51b
Chromosome Location
Chr. 10: 99107171 - 99198074 on Build GRCm38
UniGene ID
Mm.440656
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027740.6 NP_082016.1 11 1266
GenBank mRNA AK016991.1 - 10 1190
AK030180.1 - 10 1160
AK045302.1 - 11 1245
AK050233.1 - 11 1335
AK090075.1 - 11 1269
BC019688.1 - 2 151
BC050888.1 - 10 1264
BC099926.1 - 11 1352
BC144819.1 - 12 1377
BC145801.1 - 10 1274 40131104

View Full Product Details