Gene Transcripts

Gene Symbol
Poc1b
Entrez Gene ID
382406
Gene Name
POC1 centriolar protein homolog B (Chlamydomonas)
Gene Aliases
4933430F16Rik, Wdr51b
Chromosome Location
Chr. 10: 99107171 - 99198074 on Build GRCm38
UniGene ID
Mm.440656
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027740.6 NP_082016.1 10, 11 1155
GenBank mRNA AK016991.1 - 9, 10 1079
AK030180.1 - 9, 10 1049
AK045302.1 - 10, 11 1134
AK050233.1 - 10, 11 1224
AK090075.1 - 10, 11 1158
BC050888.1 - 9, 10 1153
BC099926.1 - 10, 11 1241
BC144819.1 - 11, 12 1266

View Full Product Details