Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001276448.1 | NP_001263377.1 | 4 | 373 | |
NM_177840.4 | NP_808508.2 | 4 | 373 | ||
GenBank mRNA | AK033743.1 | - | 2 | 901 | |
FJ465143.1 | - | 4 | 350 | ||
FJ465144.1 | - | 4 | 350 |